Learn More
Cayman Chemical Eicosapentaenoic Acid methyl ester, Purity: ≥98%, CAS: 2734-47-6, 50 mg
SDP
Catalog No. 502895276
Encompass_Preferred
An esterified form of EPA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-289-5276 Supplier Cayman Chemical Supplier No. 900029550 MG
An esterified form of EPA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils