Learn More
Cayman Chemical EIcosapentaenoIc ACD 1 2 3 4
SDP
Catalog No. 502988797
Encompass_Preferred
An internal standard for the quantification of EPA by GC- or LC-MS
Catalog No. 50-298-8797 Supplier Cayman Chemical Supplier No. 30017100 UG
An internal standard for the quantification of EPA by GC- or LC-MS