Learn More
Cayman Chemical EIcosapentaenoIc ACD Alkyn
SDP
Catalog No. 502966518
Encompass_Preferred
A form of EPA with an ω-terminal alkyne that can be used in click chemistry linking reactions to tag EPA with fluorescent or biotinylated labels for analysis of its metabolism and biological activity
Catalog No. 50-296-6518 Supplier Cayman Chemical Supplier No. 16704500 UG
A form of EPA with an ω-terminal alkyne that can be used in click chemistry linking reactions to tag EPA with fluorescent or biotinylated labels for analysis of its metabolism and biological activity