Learn More
Cayman Chemical EIcosapentaenoIc ACD ethyl
SDP
Catalog No. 502892236
Encompass_Preferred
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-2236 Supplier Cayman Chemical Supplier No. 9001244500 UG
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS