Learn More
Cayman Chemical EIcosapentaenoIc ACD ethyl
SDP
Supplier:  Cayman Chemical 9001244100 UG
Encompass_Preferred
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-2289