Learn More
Cayman Chemical EIcosapentaenoIc ACD ethyl50ug
SDP
Catalog No. 502892235
Encompass_Preferred
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-289-2235 Supplier Cayman Chemical Supplier No. 900124450 UG
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS