Learn More
Cayman Chemical EIcosapentaenoIc ACD methyl
SDP
Catalog No. 502892288
Encompass_Preferred
An esterified form of EPA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-289-2288 Supplier Cayman Chemical Supplier No. 9000295500 MG
An esterified form of EPA; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils