Learn More
Cayman Chemical EIcosapentaenoIc ACDd5 ethy
SDP
Catalog No. 502915743
Encompass_Preferred
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS
Catalog No. 50-291-5743 Supplier Cayman Chemical Supplier No. 37972500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of eicosapentaenoic acid ethyl ester by GC- or LC-MS