Learn More
Cayman Chemical Eicosapentaenoic Acid (peroxide free), Purity: ≥98%, CAS: 10417-94-4, 50 mg
SDP
Catalog No. 502895329
Encompass_Preferred
An ω-3 fatty acid abundantly available in marine organisms; a poor substrate for COX-1 that is oxygenated by COX-1 and COX-2 at rates of about 5% and 30%, respectively, compared to arachidonic acid; has been shown to offer protection against coronary heart disease, thrombosis, ischemic brain injury, scaly dermatitis, and some inflammatory diseases
Catalog No. 50-289-5329 Supplier Cayman Chemical Supplier No. 90110.150 MG
An ω-3 fatty acid abundantly available in marine organisms; a poor substrate for COX-1 that is oxygenated by COX-1 and COX-2 at rates of about 5% and 30%, respectively, compared to arachidonic acid; has been shown to offer protection against coronary heart disease, thrombosis, ischemic brain injury, scaly dermatitis, and some inflammatory diseases