Learn More
Cayman Chemical Eicosapentaenoic Acid, 10417-94-4, 100 mg
SDP
Supplier:  Cayman Chemical 90110100
Encompass_Preferred
Molecular Formula C20H30O2, Purity ≥98%, Formula Weight 302.5, Synonyms: EPA, Timnodonic Acid; EPA is an -3 fatty acid abundantly available in marine organisms. It is a poor substrate for COX-1.{1067} EPA has been shown to offer protection against coronary heart disease, thrombosis, ischemic brain injury, scaly dermatitis, and some inflammatory diseases.{32,208}
Catalog No. NC0458497