Learn More
Apexbio Technology LLC Eicosapentaenoic Acid(Synonyms: EPA, Timnodonic acid, Icosapentaenoic acid, all-cis-5,8,11,14,17-Eicosapentaenoic acid), 100mg, CAS: 10417-94-4.
SDP
Catalog No. 5000061059
Encompass_Preferred
Eicosapentaenoic acid (EPA CAS 10417-94-4) is an omega-3 polyunsaturated fatty acid (n-3 PUFA) extensively studied in cardiovascular research for its lipid-lowering and anti-inflammatory properties Its mechanism involves incorporation into cell membranes altering lipid composition and affecting membrane protein functions EPA inhibits endothelial cell migration and cytoskeletal rearrangements in vitro at concentrations of about 100 M Furthermore EPA (1-5 M) shows dose-dependent inhibition of very large density lipoprotein oxidation Dietary EPA enhances prostaglandin I production in human subjects suggesting protective cardiovascular effects
Catalog No. 50-000-61059 Supplier Apexbio Technology LLC Supplier No. B3464100
Eicosapentaenoic acid (EPA CAS 10417-94-4) is an omega-3 polyunsaturated fatty acid (n-3 PUFA) extensively studied in cardiovascular research for its lipid-lowering and anti-inflammatory properties Its mechanism involves incorporation into cell membranes altering lipid composition and affecting membrane protein functions EPA inhibits endothelial cell migration and cytoskeletal rearrangements in vitro at concentrations of about 100 M Furthermore EPA (1-5 M) shows dose-dependent inhibition of very large density lipoprotein oxidation Dietary EPA enhances prostaglandin I production in human subjects suggesting protective cardiovascular effects