Learn More
Medchemexpress LLC Eicosapentaenoic acid | 10417-94-4 | >99.8% | 302.45 | 250MG
SDP
Catalog No. 5000453415
Encompass_Preferred
Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It has a DNA demethylating action and promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). It can also activate the RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells and promote relaxation of vascular smooth muscle cells and vasodilation. This product is for research use only and not sold to patients.
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Promotes re-expression of tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ)
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation
  • For research use only
Catalog No. 50-004-53415 Supplier Medchemexpress LLC Supplier No. 250MGHYW011311
Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It has a DNA demethylating action and promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). It can also activate the RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells and promote relaxation of vascular smooth muscle cells and vasodilation. This product is for research use only and not sold to patients.
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Promotes re-expression of tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ)
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation
  • For research use only