Learn More
Medchemexpress LLC Eicosapentaenoic acid | 10417-94-4 | 99.86% | 302.45 | 1 G
SDP
Catalog No. 5000366565
Encompass_Preferred
Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It exhibits a DNA demethylating action that promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). This compound activates the RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells and can promote relaxation of vascular smooth muscle cells and vasodilation.
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Exhibits DNA demethylating action
  • Promotes re-expression of tumor suppressor gene
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation
Catalog No. 50-003-66565 Supplier Medchemexpress LLC Supplier No. HYW0113111G
Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It exhibits a DNA demethylating action that promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). This compound activates the RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells and can promote relaxation of vascular smooth muscle cells and vasodilation.
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Exhibits DNA demethylating action
  • Promotes re-expression of tumor suppressor gene
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation