Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eicosapentaenoic acid | 10417-94-4 | MFCD00065716 | 99.9% | 302.45 | C20H30O2 | 10 MG
SDP

Catalog No. 5000297371
Encompass_Preferred

Eicosapentaenoic acid (EPA) analytical standard supplied as a 10 mg reference material for research and analytical applications. EPA is an omega-3 long-chain polyunsaturated fatty acid commonly used as a reference in chromatographic and mass-spectrometry methods.

  • Purity 99.86%.
  • CAS number 10417-94-4.
  • Molecular formula C20H30O2; molecular weight 302.45.
  • Supplied as a 10 mg reference standard.
  • Intended for qualitative and quantitative analyses by HPLC, GC, and MS.

Catalog No. 50-002-97371 Supplier Medchemexpress LLC Supplier No. HYB0660R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eicosapentaenoic acid (EPA) analytical standard supplied as a 10 mg reference material for research and analytical applications. EPA is an omega-3 long-chain polyunsaturated fatty acid commonly used as a reference in chromatographic and mass-spectrometry methods.

  • Purity 99.86%.
  • CAS number 10417-94-4.
  • Molecular formula C20H30O2; molecular weight 302.45.
  • Supplied as a 10 mg reference standard.
  • Intended for qualitative and quantitative analyses by HPLC, GC, and MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.