Learn More
Cayman Chemical EIcosapentaenoIc AcId 250mg
SDP
Catalog No. 502895632
Encompass_Preferred
EPA is an ω-3 fatty acid abundantly available in marine organisms. It is a poor substrate for COX-1.{1067} EPA has been shown to offer protection against coronary heart disease, thrombosis, ischemic brain injury, scaly dermatitis, and some inflammatory diseases.{32,208}
Catalog No. 50-289-5632 Supplier Cayman Chemical Supplier No. 90110250 MG
EPA is an ω-3 fatty acid abundantly available in marine organisms. It is a poor substrate for COX-1.{1067} EPA has been shown to offer protection against coronary heart disease, thrombosis, ischemic brain injury, scaly dermatitis, and some inflammatory diseases.{32,208}