Learn More
Selleck Chemical LLC Eicosapentaenoic Acid
SDP
Catalog No. 5000004374
Encompass_Preferred
Eicosapentaenoic acid (EPA) is an omega-3 polyunsaturated fatty acid with antiinflammatory properties and is present in fish and fish oils including salmon cod and krill oils
Catalog No. 50-000-04374 Supplier Selleck Chemical LLC Supplier No. S647625MG
Eicosapentaenoic acid (EPA) is an omega-3 polyunsaturated fatty acid with antiinflammatory properties and is present in fish and fish oils including salmon cod and krill oils