Learn More
Medchemexpress LLC HY-B0660 50mg Medchemexpress, Eicosapentaenoic Acid CAS:10417-94-4 Purity:>98%
SDP
Catalog No. 502082635
Encompass_Preferred
Medchemexpress, HY-B0660 50mg Eicosapentaenoic Acid CAS:10417-94-4 Eicosapentaenoic Acid (EPA; Timnodonic acid) is an omega-3 fatty acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2635 Supplier Medchemexpress LLC Supplier No. HYB066050MG
Medchemexpress, HY-B0660 50mg Eicosapentaenoic Acid CAS:10417-94-4 Eicosapentaenoic Acid (EPA; Timnodonic acid) is an omega-3 fatty acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.