Learn More
Cayman Chemical Eicosapentaenoic Acid ethyl ester, Purity: ≥98%, CAS: 86227-47-6, 50 mg
SDP
Catalog No. 502889409
Encompass_Preferred
A long-chain ω-3 fatty acid ethyl ester; dietary administration increases the ex vivo activity of hepatic β-oxidation enzymes at 41.4 g/100 g of total dietary fatty acids; reduces total hepatic triglyceride levels and increases the ω-3 fatty acid content of hepatic triglycerides and phospholipids, as well as increases the total levels of ω-3 fatty acids in rats; reverses increases in plasma cholesterol levels and reduces circulating triglyceride levels in rats fed a high-fat diet at 1 g/kg
Catalog No. 50-288-9409 Supplier Cayman Chemical Supplier No. 1000888450 MG
A long-chain ω-3 fatty acid ethyl ester; dietary administration increases the ex vivo activity of hepatic β-oxidation enzymes at 41.4 g/100 g of total dietary fatty acids; reduces total hepatic triglyceride levels and increases the ω-3 fatty acid content of hepatic triglycerides and phospholipids, as well as increases the total levels of ω-3 fatty acids in rats; reverses increases in plasma cholesterol levels and reduces circulating triglyceride levels in rats fed a high-fat diet at 1 g/kg