Learn More
Medchemexpress LLC Eicosapentaenoic acid ethyl ester | 86227-47-6 | 98.0% | 500 MG
SDP
Catalog No. 5000406880
Encompass_Preferred
Eicosapentaenoic acid ethyl ester (EPA ethyl ester) is an orally active ω-3 fatty acid agent. It improves liver β-oxidase activity in vitro, reduces liver total triglyceride, and increases liver triglyceride, phospholipid ω-3 fatty acid, and total ω-3 fatty acid levels in rats. It is for research use only.
  • Orally active ω-3 fatty acid agent
  • Inhibits decrease of thyroid hormone levels and changes in thyroid follicles induced by 1-methyl-2-imidazole thiol (MMI) in hypothyroid rats
  • Reverses increased plasma cholesterol and decreased circulating triglyceride levels in high-fat diet rats
  • Related to several antibodies and small molecules
  • Supported by published research
Catalog No. 50-004-06880 Supplier Medchemexpress LLC Supplier No. HYB0747500MG
Eicosapentaenoic acid ethyl ester (EPA ethyl ester) is an orally active ω-3 fatty acid agent. It improves liver β-oxidase activity in vitro, reduces liver total triglyceride, and increases liver triglyceride, phospholipid ω-3 fatty acid, and total ω-3 fatty acid levels in rats. It is for research use only.
  • Orally active ω-3 fatty acid agent
  • Inhibits decrease of thyroid hormone levels and changes in thyroid follicles induced by 1-methyl-2-imidazole thiol (MMI) in hypothyroid rats
  • Reverses increased plasma cholesterol and decreased circulating triglyceride levels in high-fat diet rats
  • Related to several antibodies and small molecules
  • Supported by published research