Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eicosapentaenoic acid methyl ester | 2734-47-6 | 99.1% | 316.49 | 5 G
SDP

Catalog No. 5000455068
Encompass_Preferred

Eicosapentaenoic acid methyl ester is a degradant of a monogalactosyl diacylglycerol, functioning as an anticancer compound that induces apoptosis. This product is intended for research use only and is not sold to patients. It presents as a colorless to light yellow liquid with a density of 0.912 ± 0.06 g/cm3.

  • Purity: 99.11%
  • Molecular weight: 316.49
  • Chemical formula: C21H32O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Initial source: Plants such as Balsaminaceae and seeds of Impatiens balsamina L.
  • Storage for pure form: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-55068 Supplier Medchemexpress LLC Supplier No. HYW0113115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eicosapentaenoic acid methyl ester is a degradant of a monogalactosyl diacylglycerol, functioning as an anticancer compound that induces apoptosis. This product is intended for research use only and is not sold to patients. It presents as a colorless to light yellow liquid with a density of 0.912 ± 0.06 g/cm3.

  • Purity: 99.11%
  • Molecular weight: 316.49
  • Chemical formula: C21H32O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Initial source: Plants such as Balsaminaceae and seeds of Impatiens balsamina L.
  • Storage for pure form: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.