Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eicosapentaenoic Acid (Standard) | 10417-94-4 | 99.9% | 5 MG
SDP

Catalog No. 5000408468
Encompass_Preferred

Eicosapentaenoic Acid (Standard) | 10417-94-4 | 99.9% | 5 MG

Catalog No. 50-004-08468 Supplier Medchemexpress LLC Supplier No. HYB0660R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eicosapentaenoic Acid (Standard) | 10417-94-4 | 99.9% | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.