Learn More
Medchemexpress LLC Eicosapentaenoic Acid (Standard) | 10417-94-4 | 99.9% | 50 MG
SDP
Catalog No. 5000408765
Encompass_Preferred
Eicosapentaenoic Acid (Standard) is an analytical standard and orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It is used for research and analytical applications, including qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Exhibits DNA demethylating action
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-08765 Supplier Medchemexpress LLC Supplier No. HYB0660R50MG
Eicosapentaenoic Acid (Standard) is an analytical standard and orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA). It is used for research and analytical applications, including qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Orally active Omega-3 long-chain polyunsaturated fatty acid
  • Exhibits DNA demethylating action
  • Activates RAS/ERK/C/EBPβ pathway
  • Promotes relaxation of vascular smooth muscle cells and vasodilation
  • Used in HPLC, GC, and MS experiments