Learn More
Cayman Chemical Eicosapentaenoic Acidd5
SDP
Supplier:  Cayman Chemical 10005056500 UG
Encompass_Preferred
An internal standard for the quantification of EPA by GC- or LC-MS
Catalog No. 50-289-0633