Learn More
Cayman Chemical Eicosapentaenoic Acidd5
SDP
Catalog No. 502891251
Encompass_Preferred
An internal standard for the quantification of EPA by GC- or LC-MS
Catalog No. 50-289-1251 Supplier Cayman Chemical Supplier No. 100050561 MG
An internal standard for the quantification of EPA by GC- or LC-MS