Learn More
Cayman Chemical Eicosapentaenoyl Chloride, Purity: ≥95%, CAS: 98770-65-1, 50 mg
SDP
Catalog No. 502994872
Encompass_Preferred
A derivative of eicosapentaenoic acid; has been used in the synthesis of fatty acid conjugates to enhance lipophilicity and cell permeability of bioactive compounds
Catalog No. 50-299-4872 Supplier Cayman Chemical Supplier No. 2272150 MG
A derivative of eicosapentaenoic acid; has been used in the synthesis of fatty acid conjugates to enhance lipophilicity and cell permeability of bioactive compounds