Learn More
Medchemexpress LLC Eicosapentaenoyl ethanolamide | 109001-03-8 | MFCD22416528 | 98.4% | 345.52 | C22H35NO2 | 10MG
SDP
Catalog No. 5000237962
Encompass_Preferred
Eicosapentaenoyl ethanolamide is an N-acylethanolamine derived from the omega-3 fatty acid eicosapentaenoic acid. It functions as a cannabinoid CB1/CB2 receptor agonist and is supplied for research use in biochemical and pharmacological studies, with utility in assays of endocannabinoid signaling and lipid mediator pathways.
  • High purity suitable for research applications.
  • Available in small milligram pack sizes for experimental flexibility.
  • Acts as a CB1/CB2 receptor agonist in pharmacological assays.
  • Well characterized molecular identity: C22H35NO2, MW 345.52.
  • Relevant CAS identifier for compound tracking: 109001-03-8.
Catalog No. 50-002-37962 Supplier Medchemexpress LLC Supplier No. HY12408910MG
Eicosapentaenoyl ethanolamide is an N-acylethanolamine derived from the omega-3 fatty acid eicosapentaenoic acid. It functions as a cannabinoid CB1/CB2 receptor agonist and is supplied for research use in biochemical and pharmacological studies, with utility in assays of endocannabinoid signaling and lipid mediator pathways.
  • High purity suitable for research applications.
  • Available in small milligram pack sizes for experimental flexibility.
  • Acts as a CB1/CB2 receptor agonist in pharmacological assays.
  • Well characterized molecular identity: C22H35NO2, MW 345.52.
  • Relevant CAS identifier for compound tracking: 109001-03-8.