Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eicosapentaenoyl ethanolamide-d4 | 946524-41-0 | MFCD01740820 | ≥99.0% | 349.54 g·mol⁻¹ | C22H31D4NO2 | 500UG
SDP

Catalog No. 5000238424
Encompass_Preferred

Eicosapentaenoyl ethanolamide-d4 is a deuterium-labeled internal standard of eicosapentaenoyl ethanolamide intended for quantitative lipidomics and bioanalytical assays (LC-MS/MS). It contains four deuterium atoms and is provided as an analytical reference for method development and quantification in research settings.

  • Isotope-labeled internal standard for quantitative analysis.
  • Deuterium-labeled (d4) analogue optimized for mass spectrometry.
  • High purity suitable for LC-MS workflows.
  • Molecular formula C22H31D4NO2; molecular weight 349.54 g·mol⁻¹.
  • Supplied in low microgram quantities for analytical use.

Catalog No. 50-002-38424 Supplier Medchemexpress LLC Supplier No. HY124089S500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eicosapentaenoyl ethanolamide-d4 is a deuterium-labeled internal standard of eicosapentaenoyl ethanolamide intended for quantitative lipidomics and bioanalytical assays (LC-MS/MS). It contains four deuterium atoms and is provided as an analytical reference for method development and quantification in research settings.

  • Isotope-labeled internal standard for quantitative analysis.
  • Deuterium-labeled (d4) analogue optimized for mass spectrometry.
  • High purity suitable for LC-MS workflows.
  • Molecular formula C22H31D4NO2; molecular weight 349.54 g·mol⁻¹.
  • Supplied in low microgram quantities for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.