Learn More
Cayman Chemical EIcosapentaenoyl SerotonIn 1mg
SDP
Catalog No. 502893523
Encompass_Preferred
An MRGPRX2 agonist inducesBarrestin recruitment in a reporter assay using MRGPRX2-expressing HTLA cells and calcium mobilization in LAD 2 mast cells (EC50s 0.74 and 1 UM respectively) selective for MRGPRX2 over 21 other GPCRs induces degranulation of LAD 2 mast cells
Catalog No. 50-289-3523 Supplier Cayman Chemical Supplier No. 90006401 MG
An MRGPRX2 agonist inducesBarrestin recruitment in a reporter assay using MRGPRX2-expressing HTLA cells and calcium mobilization in LAD 2 mast cells (EC50s 0.74 and 1 UM respectively) selective for MRGPRX2 over 21 other GPCRs induces degranulation of LAD 2 mast cells