Learn More
Cayman Chemical EIcosapentaenoylLcarnItIn 10mg
SDP
Catalog No. 502894101
Encompass_Preferred
A long-chain acylcarnitine; levels are increased in the eyes of mice fed a diet high in n-3/n-6 PUFAs, but not in mice fed normal diet, in a mouse model of myopia induced by out-of-focus lenses
Catalog No. 50-289-4101 Supplier Cayman Chemical Supplier No. 900392410 MG
A long-chain acylcarnitine; levels are increased in the eyes of mice fed a diet high in n-3/n-6 PUFAs, but not in mice fed normal diet, in a mouse model of myopia induced by out-of-focus lenses