Learn More
Cayman Chemical EIcosapentaenoylLcarnItIn 50mg
SDP
Supplier:  Cayman Chemical 900392450 MG
Encompass_Preferred
A long-chain acylcarnitine; levels are increased in the eyes of mice fed a diet high in n-3/n-6 PUFAs, but not in mice fed normal diet, in a mouse model of myopia induced by out-of-focus lenses
Catalog No. 50-289-4099