Learn More
Cayman Chemical EIcosapentaenyl EthnlamID 50mg
SDP
Catalog No. 502951108
Encompass_Preferred
An N-acylethanolamide that inhibits dietary-restriction-induced lifespan extension in wild type and TOR pathway mutant nematodes; also has anti-inflammatory properties, suppressing the expression of IL-6 and MCP-1 in 3T3-L1 adipocytes in response to lipopolysaccharide
Catalog No. 50-295-1108 Supplier Cayman Chemical Supplier No. 1096450 MG
An N-acylethanolamide that inhibits dietary-restriction-induced lifespan extension in wild type and TOR pathway mutant nematodes; also has anti-inflammatory properties, suppressing the expression of IL-6 and MCP-1 in 3T3-L1 adipocytes in response to lipopolysaccharide