Learn More
Medchemexpress LLC HY-124108 1mg Medchemexpress, Eicosatetraynoic acid CAS:1191-85-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1241081MG
Encompass_Preferred
Medchemexpress, HY-124108 1mg Eicosatetraynoic acid CAS:1191-85-1 Eicosatetraynoic acid (ETYA) is a nonspecific inhibitor of cyclooxygenase and lipoxygenase ( ID 50 =8 μM and 4 μM, respectively) [1] . Eicosatetraynoic acid (ETYA) activates PPARα and PPARγ chimeras at 10 µM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2587