Learn More
American Radiolabeled Chemicals Inc EICOSATRIENOYL COA 1ME
SDP
Catalog No. 5090107035
Encompass
Eicosatrienoyl 8,11,14 Coenzyme A, M.W. 1055.8, Solvent: Solid in sealed ampoule under argon, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-07035 Supplier American Radiolabeled Chemicals Inc Supplier No. ARCD 02081MG
Eicosatrienoyl 8,11,14 Coenzyme A, M.W. 1055.8, Solvent: Solid in sealed ampoule under argon, Shipped in Dry Ice, Exclusive