Learn More
Aviva Systems Biology Corp Eid2 Antibody - C-terminal region (ARP53146_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eid2. It was validated on Western Blot. . Alternate names: Cri2, EID-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-801