Learn More
Enzo Life Sciences eIF-2 alpha polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPCP130D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of rat eIF2-α. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: ICC, IHC (PS), WB. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol. .
Catalog No. 50-200-7270