Learn More
Enzo Life Sciences Eukaryotic Initiation factor 2 alpha (pSer52) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4050100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of human eIF2α (eukaryotic initiation factor-2α) phosphorylated at Ser51(Ser52 in mouse). Species reactivity: Human, Mouse Yeast. Specificity: Recognizes human and yeast eIF2α phosphorylated at Ser51 and mouse eIF2α phosphorylated at Ser52. Applications: WB. Formulation: Liquid. In PBS, pH 7.3, containing 1mg/ml BSA, 50% glycerol and 0.05% sodium azide.
Catalog No. 50-201-1816