Learn More
Aviva Systems Biology Corp EIF1 Antibody - middle region (ARP63921_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63921_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF1. It was validated on Western Blot. . Alternate names: A121, EIF-1, EIF1A, ISO1, SUI1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-118