Learn More
ABclonal Technology eIF1A Rabbit mAb
SDP
Catalog No. 5000008154
Encompass_Preferred
This gene encodes an essential eukaryotic translation initiation factor The protein is required for the binding of the 43S complex (a 40S subunit eIF2 GTP Met-tRNAi and eIF3) to the 5 end of capped RNA
Catalog No. 50-000-08154 Supplier ABclonal Technology Supplier No. A2095420UL
This gene encodes an essential eukaryotic translation initiation factor The protein is required for the binding of the 43S complex (a 40S subunit eIF2 GTP Met-tRNAi and eIF3) to the 5 end of capped RNA