Learn More
Cayman Chemical eIF2α Activator 2, Purity: ≥98%, CAS: 1501618-04-7, 5 mg
SDP
Catalog No. 502943772
Encompass_Preferred
An activator of EIF2AK1/HRI; activates HRI and induces eIF2α phosphorylation in a surrogate cell-based reporter assay; selectively inhibits the proliferation of CRL-2813 cells over NIH3T3 fibroblasts (IC50s = 0.46 and 3.4 µM, respectively); selectively inhibits the proliferation of CRL-2813 cells transiently transfected with non-targeted siRNA over CRL-2813 cells transiently transfected with HRI-targeted siRNA (IC50s = 0.66 and 5 µM, respectively); increases mRNA expression and protein levels of the eIF2α target CHOP in CRL-2813 cells at 5 µM
Catalog No. 50-294-3772 Supplier Cayman Chemical Supplier No. 401305 MG
An activator of EIF2AK1/HRI; activates HRI and induces eIF2α phosphorylation in a surrogate cell-based reporter assay; selectively inhibits the proliferation of CRL-2813 cells over NIH3T3 fibroblasts (IC50s = 0.46 and 3.4 µM, respectively); selectively inhibits the proliferation of CRL-2813 cells transiently transfected with non-targeted siRNA over CRL-2813 cells transiently transfected with HRI-targeted siRNA (IC50s = 0.66 and 5 µM, respectively); increases mRNA expression and protein levels of the eIF2α target CHOP in CRL-2813 cells at 5 µM