Learn More
Aviva Systems Biology Corp EIF2A Antibody - C-terminal region (ARP41041_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41041_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human EIF2A. Alternate names: CDA02, MST089, MSTP004, MSTP089, EIF-2AApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57701