Learn More
Enzo Life Sciences eIF2alpha (human), (recombinant) (His-tag) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKPRCP1320200
Encompass_Preferred
Alternative name: Eukaryotic translation initiation factor 2 subunit alpha. Purity: ≥85% (SDS-PAGE; Western blot). MW: ~36kDa. Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 10% glycerol, 0.5% Triton X-100, 300mM potassium chloride, and 4mM DTT. Source: Produced in E. coli. Human eIF2α is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7329