Learn More
ABclonal Technology EIF2AK1 Rabbit pAb
SDP
Catalog No. 5000106519
Encompass_Preferred
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress. The encoded protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06519 Supplier ABclonal Technology Supplier No. A14119500UL
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress. The encoded protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene.