Learn More
Aviva Systems Biology Corp EIF2AK2 Antibody - middle region (ARP36709_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36709_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2AK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human EIF2AK2. Alternate names: EIF2AK1, MGC126524, PKR, PRKRApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-513