Learn More
BPS Bioscience Inc EIF2AK2 Kinase Assay Kit, 96-well plate, 100 reactions, ADP-Glo™ detection, PKR enzyme supplied, kinase substrate, ATP, assay buffer, stability 6 months
SDP
Catalog No. NC3449442
Encompass
EIF2AK2 Kinase Assay Kit, 96-well plate, 100 reactions, ADP-Glo™ detection, PKR enzyme supplied, kinase substrate, ATP, assay buffer, stability 6 months
Catalog No. NC3449442 Supplier BPS Bioscience Inc Supplier No. 78433
EIF2AK2 Kinase Assay Kit, 96-well plate, 100 reactions, ADP-Glo™ detection, PKR enzyme supplied, kinase substrate, ATP, assay buffer, stability 6 months