Learn More
Aviva Systems Biology Corp EIF2B1 Antibody - C-terminal region (ARP51963_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51963_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EIF2B1. Alternate names: EIF-2B, EIF-2Balpha, EIF2B, EIF2BA, MGC117409, MGC125868, MGC125869ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60658