Learn More
Aviva Systems Biology Corp EIF2B5 Antibody - N-terminal region (ARP61329_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61329_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2B5. It was validated on Western Blot. . Alternate names: CACH, CLE, EIF-2B, EIF2Bepsilon, LVWMApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-287-344