Learn More
Aviva Systems Biology Corp EIF2C1 Antibody - N-terminal region (ARP54863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF2C1. Alternate names: AGO1, DKFZp686M13167, EIF2C, GERP95, Q99, EIF2C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61349