Learn More
Aviva Systems Biology Corp Eif2c3 Antibody - N-terminal region (ARP58812_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58812_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eif2c3. It was validated on Western Blot. . Alternate names: AW048688, Ago3, C130014L07Rik, Eif2c3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-498