Learn More
Aviva Systems Biology Corp EIF2C3 Antibody - N-terminal region (ARP58813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2C3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF2C3. Alternate names: AGO3, FLJ12765, MGC86946, EIF2C3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62432