Learn More
Aviva Systems Biology Corp EIF2S1 Antibody - N-terminal region (ARP40507_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF2S1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF2S1. Alternate names: EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57493